Learn More
Invitrogen™ CD276 (B7-H3) Monoclonal Antibody (MJ18), FITC
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA528238
Description
This antibody recognizes mouse CD276, otherwise known as B7-H3, a ubiquitously expressed transmembrane glycoprotein and member of the B7 family of co-stimulatory molecules, which acts as both a positive and negative regulator of T-cell-mediated immune responses. CD276 is highly expressed in bone during embryogenesis, and can be induced on dendritic cells and monocytes by inflammatory cytokines.
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.
Specifications
CD276 (B7-H3) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Q8VE98 | |
CD276 | |
Mouse IgG2a Fc-CD276 (aa 1-242). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
MJ18 | |
FITC | |
CD276 | |
4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
Rat | |
Protein G | |
RUO | |
102657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.