Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD276 (B7-H3) Mouse, anti-Mouse, R718, Clone: MIH32, BD OptiBuild™

Catalog No. BDB751959 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751959 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751959 Supplier BD Biosciences Supplier No. 751959
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH32 monoclonal antibody specifically binds to CD276, also known as B7-H3 (B7 homolog 3). CD276 is a type I transmembrane glycoprotein and member of the B7-family of regulatory proteins. The expression of B7-H3 can be induced on T cells, natural killer (NK) cells and antigen presenting cells. B7-H3 is up-regulated during the differentiation of monocytes into dendritic cells or during the interaction between dendritic cells and regulatory T cells. In addition, B7-H3 is found to be expressed on fibroblasts, fibroblast-like synoviocytes and epithelial cells. CD276 (B7-H3) can function as a positive or a negative regulator of T responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD276 (B7-H3)
Applications Flow Cytometry
Classification Monoclonal
Clone MIH32
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.