Learn More
CD276 Mouse anti-Human, Brilliant Violet 510, Clone: 7-517, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743526
Description
The 7-517 monoclonal antibody specifically binds to CD276, also known as B7-H3 (B7 homolog 3). CD276 is a type I transmembrane glycoprotein and member of the B7 family of immunoregulatory proteins. Although the CD276 /gene transcript is expressed in nearly all normal tissues, the protein is preferentially expressed in tumors, tumor cell lines, sinonasal epithelial cells, and extravillous trophoblast and Hofbauer cells of the placenta. CD276 protein expression can be induced by activation of T and B lymphocytes, natural killer (NK) cells and antigen-presenting cells. The roles of CD276 in the regulation of immune responses, especially to cancer, are under investigation.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
CD276 | |
7-517 | |
Brilliant Violet 510 | |
B7-H3, B7H3, 4Ig-B7-H3, B7RP-2 | |
Human Monocyte-Derived Dendritic Cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.