Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD277/BTN3A1 Antibody (BT3.1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200227AF700
Description
CD277/BTN3A1 Monoclonal specifically detects CD277/BTN3A1 in Human samples. It is validated for Flow Cytometry.Specifications
CD277/BTN3A1 | |
Monoclonal | |
Alexa Fluor 700 | |
butyrophilin, subfamily 3, member A1, dJ45P21.3 (butyrophilin, subfamily 3, member A1) | |
Mouse | |
Affinity Purified | |
RUO | |
11119 | |
Store at 4C in the dark. |
Flow Cytometry | |
BT3.1 | |
Flow Cytometry | |
BTN3A1 | |
The immunogen for this antibody is CD277 (B7 family, Butyrophilin). | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction