Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD277/BTN3A1 Antibody (BT3.1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200227B
Description
CD277/BTN3A1 Monoclonal antibody specifically detects CD277/BTN3A1 in Human samples. It is validated for Flow Cytometry, ImmunoprecipitationSpecifications
CD277/BTN3A1 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
11119 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
BT3.1 | |
Flow Cytometry, Immunoprecipitation | |
butyrophilin, subfamily 3, member A1, dJ45P21.3 (butyrophilin, subfamily 3, member A1) | |
The immunogen for this antibody is CD277 (B7 family, Butyrophilin). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction