Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD278 (ICOS) Monoclonal Antibody (C398.4A), APC, eBioscience™, Invitrogen™

Catalog No. 17994980
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-994-980 25 μg
17-994-982 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-994-980 Supplier Invitrogen™ Supplier No. 17994980
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: The C398.4A monoclonal antibody reacts with ICOS (Inducible Costimulatory molecule), also known as H4/CRP-1/AILIM, and cross-reacts with mouse and rat ICOS. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. A dimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1, is thought to play a protective role in inflammatory autoimmune diseases and be involved in the development of Th2 cells. C398.4A is reported to display strong costimulation of proliferation of T cell clones, intermediate costimulation on activated T cells, and weak costimulation on fresh resting T cells. It is a good costimulator of IL-10 production especially from pre-activated cells that express high level of H4/ICOS. The epitope recognized by C398.4A is different from that bound by the ICOSL (i.e., C398.4A does not block binding of ICOSL). Applications Reported: This C398.4A antibody has been reported for use in flow cytometric analysis. Applications Tested: This C398.4A antibody has been tested by flow cytometric analysis of Con A-stimulated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD278 (ICOS)
Applications Flow Cytometry
Classification Monoclonal
Clone C398.4A
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ICOS
Gene Accession No. Q9R1T7, Q9WVS0
Gene Alias Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115
Gene Symbols ICOS
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54167, 64545, 705788
Target Species Mouse, Rat, Rhesus Macaque
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.