Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD278 (ICOS) Monoclonal Antibody (ISA-3), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ H060T03Y07A

Description
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
CD278 (ICOS) | |
Monoclonal | |
0.6 μg/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9Y6W8 | |
ICOS | |
Affinity chromatography | |
RUO | |
29851 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid | |
IgG1 κ |
Flow Cytometry | |
ISA-3 | |
NovaFluor Yellow 730 | |
ICOS | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction