Learn More
Invitrogen™ CD278 (ICOS) Monoclonal Antibody (ISA-3), Super Bright™ 436, eBioscience™
Mouse Monoclonal Antibody
$293.65
Specifications
Antigen | CD278 (ICOS) |
---|---|
Clone | ISA-3 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
62-994-842
![]() |
Invitrogen™
62994842 |
100 Tests |
Each of 1
for $293.65
Save 25% Reg : |
![]() |
|||||
Description
Description: The ISA-3 monoclonal antibody reacts with human CD278 also known as Inducible Costimulatory molecule (ICOS), H4, CRP-1 and AILIM. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. Human ICOS has a relative molecular mass of 55-60 kDa, composed of 27 kDa and 29 kDa chains. Human ICOS on activated T cells has potent costimulatory activity for T cell activation and is required for humoral immune responses, in particular for memory B cell and plasma cell generation. ICOS binds to its ligand, B7h/B7RP-1 expressed on activated APCs (antigen presenting cells) and on a number of inflamed peripheral tissues. Plate-bound ISA-3 is costimulatory for T cells and induces production of IL-4, IL-5, IL-10 and other cytokines, but not IL-2. ISA-3 has the same reactivity pattern and characteristics as F44. ISA-3 was generated against the human ICOS antigen. C398.4A, anti-mouse ICOS/H4 (Product # 14-9949-82) , was shown to cross-react with human ICOS but binds to an epitope different from ISA-3. C398.4A stains activated cells brighter than ISA-3; however, it also exhibits higher staining of non-activated human peripheral blood or isolated PBMC. To achieve the brightest staining of ICOS on activated human T cells, please use Product # 13-9948-82 or Product # 12-9948-42 rather than Product # 11-9948-42 . Applications Reported: This ISA-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ISA-3 antibody...
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.Specifications
CD278 (ICOS) | |
5 μL/Test | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q9Y6W8 | |
29851 | |
Primary | |
Affinity Chromatography |
ISA-3 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
ICOS | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
ICOS | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.