Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 Hamster anti-Mouse, BUV395, Clone: J43, BD Optibuild™

Catalog No. BDB744549 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744549 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744549 Supplier BD Biosciences Supplier No. 744549
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The J43 monoclonal antibody specifically recognizes CD279 which is also known as PD-1 (programmed death-1). CD279 is a 50-55-kDa glycoprotein encoded by the Pdcd1 /gene of the CD28 family of the Ig superfamily. The expression of Pdcd1 mRNA and PD-1 protein is tightly regulated. PD-1 is transiently expressed on CD4-CD8 thymocytes, it is upregulated on some cell lines upon induction of apoptosis, it is induced on thymocytes and splenic T and B lymphocytes after stimulation through their antigen receptors, and it is induced on activated myeloid cells. In addition, Pdcd1 /mRNA is transiently expressed in developing B lymphocytes at the pro-B-cell stage. The presence of an ITIM ( I /mmunoreceptor T /yrosine-based I /nhibitory M /otif) on PD-1's intracytoplasmic region and the development of splenomegaly and breakdown of peripheral tolerance in PD-1[-/-] mice suggest that PD-1 may be involved in the negative regulation of immune responses. The PD-1 ligands, B7-H1 (also known as PD-L1) and B7-DC (PD-L2), are members of the B7 family of the Ig superfamily. The J43 antibody blocks the binding of PD-1 to its two ligands.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279
Classification Monoclonal
Clone J43
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PD-1
Host Species Armenian Hamster
Immunogen Syrian Hamster kidney cell line BKH transfected with Pdcd1 cDNA
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.