Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 Mouse anti-Human, FITC, Clone: EH12.2H7, Proteintech
SDP

Catalog No. 502258294 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-8294 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-8294 Supplier Proteintech Group Inc Supplier No. FITC65168100TESTS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD279 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279. This antibody recognizes non-human primates, human antigen. The CD279 antibody has been validated for the following applications: FC analysis. This antibody is fitc.

Specifications

Antigen CD279
Applications Flow Cytometry
Classification Monoclonal
Clone EH12.2H7
Concentration 4 μL/Test
Conjugate FITC
Host Species Mouse
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human, Non-human Primate
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.