Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 Mouse anti-Human, PE, Clone: MIH4, BD

Catalog No. BDB560908 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560908 25 Tests
BDB557946 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560908 Supplier BD Biosciences Supplier No. 560908
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH4 monoclonal antibody specifically binds to the program death 1 (PD-1) receptor CD279, a member of the Ig superfamily. CD279 is an immunoregulatory receptor expressed on activated T cells, B cells and myeloid cells. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. It contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279
Applications Flow Cytometry
Classification Monoclonal
Clone MIH4
Conjugate PE
Description PD-1; hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PD-1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.