Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Monoclonal Antibody (J43), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$215.00 - $437.00
Specifications
Antigen | CD279 (PD-1) |
---|---|
Clone | J43 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
37-699-8582
![]() |
Invitrogen™
376998582 |
100 μg |
Each for $437.00
|
|
|||||
37-699-8580
![]() |
Invitrogen™
376998580 |
25 μg |
Each for $215.00
|
|
|||||
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow CytometrySpecifications
CD279 (PD-1) | |
0.2 mg/mL | |
Monoclonal | |
Armenian Hamster | |
Mouse | |
Q02242 | |
18566 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
J43 | |
Flow Cytometry | |
Brilliant Ultraviolet 661 | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
IgG | |
Affinity chromatography | |
Pdcd1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title