Learn More
CD279 (PD-1) Monoclonal Antibody (J43), eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$210.00 - $384.00
Specifications
Antigen | CD279 (PD-1) |
---|---|
Clone | J43 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-139-66
![]() |
Invitrogen™
14998582 |
100 μg |
Each for $210.00
|
![]() |
|||||
50-112-9003
![]() |
Invitrogen™
14998585 |
500 μg |
Each for $384.00
|
![]() |
|||||
Description
Description: The J43 monoclonal antibody reacts with mouse PD-1 (programmed death-1), a 55 kDa member of the Ig superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease in mice. PD-1 is expressed mainly on activated T and B lymphocytes. Two novel B7 Family members have been identified as PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. It is reported that J43 inhibits the binding of mouse PD-L1-Ig and mouse PD-L2-Ig to PD-1/BHK transfected cells. When administrated in vivo, both intact and Fab of J43 are reported to enhance contact hypersensitivity and exacerbate acute GVHD similar to transfer of PD-1-deficient cells. Injection of J43 also exacerbates EAE and NOD diabetes as do specific antibodies to mouse PD-L1 and PD-L2. Applications Reported: The J43 antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. It has also been reported for use in in vitro functional assays. (Please use Functional Grade purified J43, cat. 16-9985, in functional assays.). Applications Tested: The J43 antibody has been tested by flow cytometric analysis of mouse Con-A activated spleen cells and mouse PD-1 transfected cells. This can be used at less than or equal to 0.
Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "e;danger signals"e; such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.Specifications
CD279 (PD-1) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q02242 | |
Pdcd1 | |
Primary | |
4°C | |
Pdcd1 |
J43 | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
Unconjugated | |
Armenian Hamster | |
Mouse | |
18566 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.