Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 (PD-1) Mouse anti-Human, BB515, Clone: EH12.1, BD Horizon™

Catalog No. BDB564494 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564494 50 Tests
BDB565936 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB564494 Supplier BD Biosciences Supplier No. 564494
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279 (PD-1)
Applications Flow Cytometry
Classification Monoclonal
Clone EH12.1
Conjugate Brilliant Blue 515 (BB515)
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2
Host Species Mouse
Immunogen Human PD-1 Recombinant Protein
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.