Learn More
CD279 (PD-1) Mouse anti-Human , BUV615, Clone: EH12.1 (also known as EH12), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD279 (PD-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | EH12.1 (also known as EH12) |
| Conjugate | Brilliant Ultraviolet 615 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2; hSLE1 |
| Host Species | Mouse |
| Immunogen | Human PD-1 Recombinant Protein |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.