Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human , PE-CF594, Clone: MIH4, BD Biosciences™
Mouse Monoclonal Antibody
$174.70 - $416.00
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | MIH4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE-CF594 |
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow CytometrySpecifications
| CD279 (PD-1) | |
| Flow Cytometry | |
| PE-CF594 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human PD-1 Transfected Cell Line | |
| Primary |
| MIH4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title