Learn More
CD279 (PD-1) Mouse anti-Human, PE, Clone: J110, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc PE65119100TESTS
Description
The CD279 (PD-1) antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279 (PD-1). This antibody recognizes human antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
CD279 (PD-1) | |
Monoclonal | |
5 μL (1 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
J110 | |
PE | |
100 Tests | |
5133 | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.