Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 (PD-1), Mouse Anti-Human, RB613, EH12.1 (also known as EH12), BD Horizon™

Catalog No. BDB571094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571094 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB571094 Supplier BD Biosciences Supplier No. 571094
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human (QC Testing) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279 (PD-1)
Applications Flow Cytometry
Classification Monoclonal
Clone EH12.1 (also known as EH12)
Conjugate RB613
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2; hSLE1
Host Species Mouse
Immunogen Human PD-1 Recombinant Protein
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.