Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Rat anti-Mouse, BUV615, Clone: RMP1-30, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 752354
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow Cytometry.

Specifications
CD279 (PD-1) | |
Monoclonal | |
0.2 mg/mL | |
Ms CD279 (PD-1) BUV615 RMP1-30 50 μg | |
Mouse PD-1 Recombinant Protein | |
RUO | |
CD279 (PD-1) | |
Refrigerate (2 to 8°C) | |
Mouse |
Flow Cytometry | |
RMP1-30 | |
Brilliant Ultraviolet 615 | |
Rat | |
1 x 50 μg | |
Primary | |
Mouse | |
IgG2b κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction