Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 (PD-1) Rat anti-Mouse, BV650, Clone: 29F.1A12, BD Horizon™

Rat Monoclonal Antibody

$104.50 - $253.50

Specifications

Antigen CD279 (PD-1)
Clone 29F.1A12
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB568616
View Documents
BD Biosciences
568616
0.1 mg
Each for $253.50
Only null left
Add to Cart
 
BDB568617
View Documents
BD Biosciences
568617
25 μg
Each for $104.50
Only null left
Add to Cart
 
Description

Description

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow cytometry
Specifications

Specifications

CD279 (PD-1)
0.2 mg/mL
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
PD-1 cDNA and PD-1-Ig Fusion Protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
29F.1A12
Flow Cytometry
Brilliant Violet 650
Rat
Mouse
PD-1; Pdc1; Pdcd1; mPD-1; programmed cell death 1 protein
IgG2a κ
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.