Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Rat anti-Mouse, Clone: 29F.1A12, BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568262
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Blocking, Flow Cytometry
Specifications
CD279 (PD-1) | |
Monoclonal | |
1.0 mg/mL | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Rat | |
2 mg | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. | |
IgG2a κ |
Blocking Assay, Flow Cytometry | |
29F.1A12 | |
Unconjugated | |
PD-1; Pdc1; Pdcd1; mPD-1; programmed cell death 1 protein | |
PD-1 cDNA and PD-1-Ig Fusion Protein | |
RUO | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction