Learn More
CD279 (PD-1) Rat anti-Mouse , PE-CF594, Clone: RMP1-30, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD279 (PD-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RMP1-30 |
| Conjugate | PE-CF594 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | PD-1; mPD-1; Pdc1; Pdcd1; Ly101 |
| Host Species | Rat |
| Immunogen | Mouse PD-1 Recombinant Protein |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.