Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Rat anti-nan, BV510, Clone: 29F.1A12, BD Horizon™
Rat Monoclonal Antibody
$130.00
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | 29F.1A12 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in nan samples. It is validated for Flow cytometrySpecifications
| CD279 (PD-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PD-1; Pdc1; Pdcd1; mPD-1; programmed cell death 1 protein | |
| IgG2a κ | |
| Affinity Chromatography |
| 29F.1A12 | |
| Flow Cytometry | |
| Brilliant Violet 510 | |
| Rat | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| PD-1 cDNA and PD-1-Ig Fusion Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title