Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Antibody (204.12), Novus Biologicals™

Mouse Monoclonal Antibody
$281.00 - $524.00
Specifications
| Antigen | CD28 |
|---|---|
| Clone | 204-12 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24476000
![]() |
Novus Biologicals
NBP2447600.02MG |
0.02 mg |
Each for $281.00
|
|
|||||
NBP24476001
![]() |
Novus Biologicals
NBP2447600.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24476002
![]() |
Novus Biologicals
NBP2447600.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD28 Monoclonal specifically detects CD28 in Human, Mouse, Porcine, Bovine, Sheep samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.Specifications
| CD28 | |
| 0.2mg/mL | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 940 | |
| Stimulated human leucocytes | |
| Primary | |
| Store at 4C. |
| 204-12 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig, Primate, Sheep | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| CD28 | |
| IgG2a κ | |
| Protein A or G purified | |
| Recognizes a glycoprotein of 44-88kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title