Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Antibody (CD28.2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179126H
Description
CD28 Monoclonal specifically detects CD28 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD28 | |
Monoclonal | |
HRP | |
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody CD28.2 reacts with CD28, a disulfide-linked homodimeric type I glycoprotein (monomer of Mw 44 kDa) which is a critical costimulatory receptor of T cells. HLDA V.; WS Code 5T CD28.05 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CD28.2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD28 | |
DC28.1.3.3 murine T cell hybridoma transfected with human CD28 cDNA | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
940 | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction