Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Antibody (JJ316), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281052PCP
Description
CD28 Monoclonal specifically detects CD28 in Rat samples. It is validated for Flow Cytometry.Specifications
CD28 | |
Monoclonal | |
PerCP | |
PBS | |
CD28 | |
Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells. | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
940 | |
Rat | |
Purified |
Flow Cytometry | |
JJ316 | |
Flow Cytometry | |
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises rat CD28 and can trigger T-cell proliferation in vitro and in vivo. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction