Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Antibody (JJ319) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200000
Description
CD28 Monoclonal specifically detects CD28 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD28 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Rat | |
IgG1 |
Flow Cytometry | |
JJ319 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
CD28 | |
The immunogen for this antibody is CD28. | |
0.5 mg | |
Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
940 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction