Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Hamster anti-Mouse, Clone: 37.51, BD Pharmingen™

Catalog No. BDB567110 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567110 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567110 Supplier BD Biosciences Supplier No. 567110
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hamster Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry-frozen, (Co)-stimulation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 37.51
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing protein stabilizer, no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay.
Gene Alias Cd28; CD28 antigen; T-cell-specific surface glycoprotein CD28
Host Species Hamster
Immunogen Mouse EL-4 (T-cell lymphoma) Cells
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions.
Form Liquid
Isotype IgG2 λ1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.