Learn More
Invitrogen™ CD28 Monoclonal Antibody (B-23), FITC

Description
CD28 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD28 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B-23 |
| Concentration | 30 μg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.2% carrier protein and 0.08% sodium azide |
| Gene | Cd28 |
| Gene Accession No. | P10747 |
| Gene Alias | antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.