Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Monoclonal Antibody (JJ319), FITC
CD28 Monoclonal Antibody (JJ319), FITC
Supplier: Thermo Scientific MA517471
Description
Specifications
| CD28 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P31042 | |
| Cd28 | |
| Rat CD28 transfected A20J cells. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| JJ319 | |
| FITC | |
| Cd28 | |
| antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
| Mouse | |
| Protein G | |
| RUO | |
| 25660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction