Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, PE, Clone: CD28.2, Abnova™

Catalog No. 89933768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-768 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-768 Supplier Abnova Corporation Supplier No. MAB12224
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD28.

CD28 costimulation is essential for CD4 (MIM 186940)-positive T-cell proliferation, survival, interleukin-2 (IL2; MIM 147680) production, and T-helper type-2 (Th2) development.[supplied by OMIM]

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone CD28.2
Conjugate PE
Description Mouse monoclonal antibody raised against CD28.
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer).
Gene Alias MGC138290/Tp44
Gene Symbols CD28
Host Species Mouse
Immunogen Native purified CD28 from DC28.1.3.3 hybridoma cells.
Purification Method Affinity Chromatography
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.