Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, Clone: CD28.2, BD Pharmingen™

Catalog No. BDB567117 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567117 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567117 Supplier BD Biosciences Supplier No. 567117
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow Cytometry, Activation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone CD28.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay.
Gene Alias CD28 antigen; T44; Tp44; TP44
Host Species Mouse
Immunogen Human CD28 Transfected Cell Line
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Macaque, Cynomolgus, Baboon
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.