Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28, Mouse Anti-Human, RY610, Clone: CD28.2, BD Horizon™

Catalog No. BDB571178 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Conjugate:
RY610
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571178 25 Tests RY610
BDB571177 100 Tests RY610
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571178 Supplier BD Biosciences Supplier No. 571178
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone CD28.2
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD28 antigen; T44; Tp44; TP44
Host Species Mouse
Immunogen Human CD28 Transfected Cell Line
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940, 705313, 100126740, 102146670
Target Species Human, Rhesus Monkey, Cynomolgus, Baboon
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.