Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse Anti-Rat, BUV395, Clone: JJ319, BD Optibuild™

Catalog No. BDB742588 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742588 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742588 Supplier BD Biosciences Supplier No. 742588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JJ319 monoclonal antibody specifically recognizes CD28, which is expressed on virtually all T lymphocytes bearingαβT-cell receptors (TCR), on mostγδTCR-bearing T cells, and on a subset of NK cells. In the thymus, CD28 expression is developmentally regulated during the maturation ofαβTCR-bearing T cells. CD28 is a costimulatory receptor required for activation of T cells; its ligands include CD80 (B7-1) and CD86 (B7-2). Soluble JJ319 mAb costimulates the proliferative responses and IL-2 production of CD4+ and CD8+ T cells activated by anti-αβ-TCR mAb R73 (Cat. no. 554910). The alternate anti-rat CD28 mAb JJ316 (Cat. No. 554992) is capable of directly stimulating T cells in vitro and in vivo.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Classification Monoclonal
Clone JJ319
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd28, CD28RNA
Host Species Mouse
Immunogen Rat CD28-transfected cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.