Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Syrian Hamster anti-Mouse, Unconjugated, Clone: 37.51, MilliporeSigma™

Catalog No. MABF208925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2089-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2089-25 Supplier MilliporeSigma Supplier No. MABF208925UG
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

This syrian hamster monoclonal Anti-CD28, clone 37.51 Antibody, Cat. No. MABF2089 is tested for use in Flow Cytometry, Western Blotting, Immunoprecipitation, and Affects Function, for the detection of CD28.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry, Western Blot, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone 37.51
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified syrian hamster monoclonal antibody IgG2-IgG3 in PBS without azide.
Gene Alias T-cell-specific surface glycoprotein CD28
Gene Symbols Cd28
Host Species Syrian Hamster
Immunogen EL-4 T lymphoma cells.
Molecular Weight of Antigen 25.24 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_031668.3
Test Specificity Clone 37.51 is a Syrian hamster monoclonal antibody that detects CD28 in murine cells.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.