Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD282 (TLR2) Mouse anti-Human, Brilliant Violet 711, Clone: 11G7, BD Optibuild™

Catalog No. BDB742770 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742770 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742770 Supplier BD Biosciences Supplier No. 742770
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The 11G7 monoclonal antibody specifically binds to human CD282, which is also known as Toll-like receptor 2 (TLR2). CD282 is expressed on monocytes, granulocytes, and dendritic cells. Toll-like receptors (TLRs) play a critical role in antimicrobial resistance. Moreover, TLRs have been shown to activate a number of signal transduction pathways which lead to the induction of genes involved in host defense. TLRs are type-1 transmembrane receptors characterized by the presence of extracellular leucine-rich repeat and intracellular Toll/IL-1 receptor domains. At least 12 mammalian TLRs have been identified, each recognizing a distinct bacterial or viral pathogen-associated molecular pattern, termed PAMP. Peptidoglycan from Gram-positive bacteria, lipoproteins and lipopeptides from several bacteria, glycophosphatidylinositol, lipoarabinomannan, porins, and zymosan from yeast have been reported to be the ligands for TLR2. It has been reported that mAb 11G7 inhibits the production of inflammatory cytokines via certain TLR2 ligands including TLR2/TLR1 ligands, lipoarabinomannan and PAM3CSK4. However, 11G7 antibody does not inhibit the production of inflammatory cytokines with zymosan, a TLR2/TLR6 ligand. Please note that this application has not been tested at BD Biosciences Pharmingen.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD282 (TLR2)
Classification Monoclonal
Clone 11G7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TLR2, CD282, TIL4, Toll/interleukin-1 receptor-like protein 4
Host Species Mouse
Immunogen Human TLR2-transfected cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.