Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD282 (TLR2) Mouse anti-Human, BUV395, Clone: 11G7, BD Optibuild™

Catalog No. BDB742772 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742772 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742772 Supplier BD Biosciences Supplier No. 742772
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11G7 monoclonal antibody specifically binds to human CD282, which is also known as Toll-like receptor 2 (TLR2). CD282 is expressed on monocytes, granulocytes, and dendritic cells. Toll-like receptors (TLRs) play a critical role in antimicrobial resistance. Moreover, TLRs have been shown to activate a number of signal transduction pathways which lead to the induction of genes involved in host defense. TLRs are type-1 transmembrane receptors characterized by the presence of extracellular leucine-rich repeat and intracellular Toll/IL-1 receptor domains. At least 12 mammalian TLRs have been identified, each recognizing a distinct bacterial or viral pathogen-associated molecular pattern, termed PAMP. Peptidoglycan from Gram-positive bacteria, lipoproteins and lipopeptides from several bacteria, glycophosphatidylinositol, lipoarabinomannan, porins, and zymosan from yeast have been reported to be the ligands for TLR2. It has been reported that mAb 11G7 inhibits the production of inflammatory cytokines via certain TLR2 ligands including TLR2/TLR1 ligands, lipoarabinomannan and PAM3CSK4. However, 11G7 antibody does not inhibit the production of inflammatory cytokines with zymosan, a TLR2/TLR6 ligand. Please note that this application has not been tested at BD Biosciences Pharmingen.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD282 (TLR2)
Classification Monoclonal
Clone 11G7
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TLR2, CD282, TIL4, Toll/interleukin-1 receptor-like protein 4
Host Species Mouse
Immunogen Human TLR2-transfected cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.