Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD283 (TLR3) Monoclonal Antibody (TLR3.7), Functional Grade, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 16903982

Encompass_Preferred

Catalog No. 50-147-76

  • $414.00
    $310.65 / Each of 1

    Save $103.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart

Description

Description

Description: The TLR3.7 monoclonal antibody reacts with human Toll-like receptor 3 (TLR3). To date, at least twelve members of the Toll-like receptor family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. In the innate immune response, TLRs recognize molecular patterns associated with microbial pathogens and induce antimicrobial activity. TLR3 recognizes double-stranded (ds)RNA, induces the activation of NF-kappaB through MyD88-dependent and -independent pathways, and the production of type I interferons (IFNs). TLR3.7 suppressed poly(I):poly(C)-mediated IFN-beta production by human fibroblasts naturally expressing TLR3 on their surface. Applications Reported:The TLR3.7 antibody has been reported for use in flow cytometric analysis. It has also been reported in in vitro inhibition of ligand binding. For optimal detection of TLR3 with this mAb by flow cytometry, PE-conjugated or Biotin-conjugated TLR3.7 (revealed with SAv-PE) are recommended. Applications Tested: This TLR3.7 antibody has been tested by intracellular staining and flow cytometric analysis of A549 and MRC5 cell lines to detect the very low level of this antigen on the surface. This can be used at less than or equal to 2 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell num...

TLR3 (Toll-like receptor 3, CD283) is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from drosophila to humans and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR3 is most abundantly expressed in placenta and pancreas, and is restricted to the dendritic subpopulation of the leukocytes. TLR3 recognizes dsRNA associated with viral infection, and induces the activation of NF-kappaB and the production of type I interferons. Further, TLR3 may play a role in host defense against viruses, and the use of alternative polyadenylation sites to generate different length transcripts has been noted for the TLR3 gene. Like its counterparts in Drosophila, TLRs signal through adaptor molecules and could constitute an important and unrecognized component of innate immunity in humans. The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition. TLRs characterized so far activate the MyD88/interleukin-1 receptor-associated kinase (IRAK) signaling pathway. TLR3 is the only human TLR which does not utilize MyD88 as an adaptor molecule instead utilizing TRIF in a signaling path which results in downstream activation of IRF-3 and NF-kB. TLR3 activation leads not only to Type I interferon induction but also to other inflammatory cytokines which result in DC maturation. TLR3 is known to recognize viral double-stranded (ds) RNA, a molecular pattern associated with viral infection. Recently it has been shown to recognize viruses such as Influenza A and West Nile Virus and can mediate entry of at least West Nile Virus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD283 (TLR3)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
O15455
TLR3
Affinity chromatography
RUO
7098
4°C
Liquid
Flow Cytometry, Functional Assay, Neutralization
TLR3.7
Functional Grade
TLR3
AI957183; CD283; IIAE2; TLR 3; TLR3; TLR-3; toll like receptor 3; toll-like receptor 3; toll-like receptor 3 variant 1
Mouse
100 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.