Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD284/MD-2 Complex Rat anti-Mouse, Oligo, Clone: MTS510, BD™ AbSeq ™

Catalog No. BDB940457 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB940457 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB940457 Supplier BD Biosciences Supplier No. 940457
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD284/MD-2 Complex Monoclonal antibody specifically detects CD284/MD-2 Complex in Mouse samples. It is validated for Single Cell 3' Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD284/MD-2 Complex
Applications Single Cell 3' Sequencing
Classification Monoclonal
Clone Clone MTS510
Conjugate Oligo
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias TLR4/MD-2 Complex
Host Species Rat
Immunogen Mouse Pro-B cell line Ba/F3 expressing transfected BALB/c mouse TLR4 and MD-2
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.