Learn More
Invitrogen™ CD284 (TLR4) Monoclonal Antibody (HTA125), APC-eFluor™ 780, eBioscience™, Invitrogen™

Description
Specifications
Specifications
| Antigen | CD284 (TLR4) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HTA125 |
| Concentration | 5 μL/Test |
| Conjugate | APC-eFluor 780 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | TLR4 |
| Gene Accession No. | O00206 |
| Gene Alias | ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.