Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD284 (TLR4) Mouse anti-Human, Brilliant Violet 421, Clone: HTA125, BD Optibuild™

Catalog No. BDB743392 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743392 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743392 Supplier BD Biosciences Supplier No. 743392
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HTA125 monoclonal antibody specifically binds to the human Toll-like receptor 4 (TLR4), also known as CD284. Toll-like receptors (TLR) are type I transmembrane proteins characterized by an extracellular domain containing leucine-rich repeats and a cytoplasmic domain similar to IL-1R family. In humans, ten TLR have been identified so far. The TLR family acts as a pattern-recognition receptor that is an essential component of pathogen recognition and innate immunity. TLR4 has been identified as the receptor for Gram-negative bacterial lipopolysaccharide (LPS). TLR4 mRNA expression has been found in spleen, placenta, ovary, intestine and lung. Using HTA125, TLR4 protein expression has been detected in monocytes, B cells and T cells in human peripheral blood. The human TLR4 /gene has been mapped to chromosome 9q32. The immunogen used to generate the HTA125 hybridoma was human TLR4-transfected Ba/F3 cells. The HTA125 antibody reportedly blocks LPS-triggered, TLR4-mediated stimulation of cells.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD284 (TLR4)
Classification Monoclonal
Clone HTA125
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TLR4, CD284, Toll-like Receptor 4, ARMD10
Host Species Mouse
Immunogen Human TLR4-transfected Ba/F3 cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.