Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD286 (TLR6) Mouse anti-Mouse, Brilliant Violet 786, Clone: C1N2, BD Optibuild™

Catalog No. BDB743271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743271 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743271 Supplier BD Biosciences Supplier No. 743271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C1N2 monoclonal antibody specifically binds to the Toll-like receptor 6 (TLR6), which is also known as CD286. CD286 belongs to the mammalian toll-like family of pattern recognition receptors (TLR) that participate in inducing innate immune responses to microbial pathogens. CD286 is a type I transmembrane glycoprotein comprised of an extracellular domain with a large number of leucine?rich repeats (LRRs) and a cytoplasmic region with a signaling Toll/Interleukin-1 Receptor (TIR) domain. TLR6 is differentially expressed on the surface and intracellular compartments, including endolysosomes, of pre-B cells as well as immature or mature B cells. It is likewise expressed by monocytes, macrophages, or plasmacytoid, CD4+, or CD8+ dendritic cells. In association with TLR2 (CD282), CD286 recognizes microbial diacylated lipopeptides, lipoteichoic acid, and zymosan. The CD282/CD286 (TLR2/TLR60 heterodimer then transduces signaling responses leading to the production of proinflammatory cytokines by responding cells.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD286 (TLR6)
Classification Monoclonal
Clone C1N2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Tlr6, Toll-like receptor 6
Host Species Mouse
Immunogen Mouse TLR6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.