Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD29 (Integrin β1 chain) Armenian Hamster anti-Mouse, Oligo, Clone: HM β1-1, BD™ AbSeq ™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 940158
This item is not returnable.
View return policy
Description
CD29 (Integrin β1 chain) Monoclonal antibody specifically detects CD29 (Integrin β1 chain) in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
CD29 (Integrin β1 chain) | |
Monoclonal | |
Oligo | |
Itgb1; Integrin beta-1; Fnrb; gpIIa; VLA-4 subunit beta | |
Purified mouse VLA-4 | |
RUO | |
Mouse | |
Purified |
Single Cell 3' Sequencing | |
Clone HM β1-1 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Armenian Hamster | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. | |
IgG2 λ1 |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction