Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD29 (Integrin beta 1) Monoclonal Antibody (eBioHMb1-1 (HMb1-1)), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | CD29 (Integrin beta 1) |
---|---|
Clone | eBioHMb1-1 (HMb1-1) |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M048T03Y06A
![]() |
Invitrogen™
M048T03Y06A |
100 μg |
Each for $379.00
|
|
|||||
M048T02Y06A
![]() |
Invitrogen™
M048T02Y06A |
25 μg |
Each for $201.00
|
|
|||||
Description
CD29 (Integrin beta 1) Monoclonal antibody specifically detects CD29 (Integrin beta 1) in Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CD29 (Integrin beta 1) | |
0.1 mg/mL | |
Monoclonal | |
Armenian Hamster | |
Mouse, Rat | |
P09055, P49134 | |
16412, 24511 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioHMb1-1 (HMb1-1) | |
Flow Cytometry | |
NovaFluor Yellow 700 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
IgG | |
Affinity chromatography | |
ITGB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title