Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD29 Mouse anti-Human, BV421, Clone: TS2/16, BD Horizon™
Mouse Monoclonal Antibody
$168.70 - $414.00
Specifications
Antigen | CD29 |
---|---|
Clone | TS2/16 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
CD29 Monoclonal antibody specifically detects CD29 in Human samples. It is validated for Flow cytometrySpecifications
CD29 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
FNRB; GPIIA; ITGB1; MDF2; MSK12; VLA-BETA; VLAB; glycoprotein IIa | |
Human CTL Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
TS2/16 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3688 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title