Learn More
Invitrogen™ CD294 (CRTH2) Monoclonal Antibody (BM16), Biotin, eBioscience™
Rat Monoclonal Antibody
$169.00
Specifications
Antigen | CD294 (CRTH2) |
---|---|
Clone | BM16 |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
13-294-982
![]() |
Invitrogen™
13294982 |
100 μg |
Each of 1 for $169.00
|
![]() |
|||||
Description
Description: This BM16 monoclonal antibody reacts with human CD294, which is also known as G protein-coupled receptor-44 (GPR-44) or chemoattractant receptor expressed on Th2 cells (CRTH2). CD294 is expressed on Th2 cells, some CD8+ cells, eosinophils, and basophils. Moreover, CD294 binds prostaglandin D2, which promotes leukocyte chemotaxis during allergic responses. CD294 has been reported to mediate prostaglandin D2-mediated suppression of apoptosis of Th2 cells by activating the PI3K pathway. Applications Reported: This BM16 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BM16 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CRTH2 (GPR44) is a chemoattractant receptor-homologous molecule expressed on Th2 cells that is an identified receptor for the prostaglandin D2 (PGD2) and prostaglandin D receptor. CRTH2 is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, CRTH2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the CRTH2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that CRTH2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo. CRTH2 is also a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells. CRTH2 has been reported to be expressed in the central nervous system, peripheral tissues, and in blood cells, particularly activated T helper-2 cells. Two transcripts have been isolated: a 3.5 kb transcript expressed primarily in the thalamus, frontal cortex, pons and hippocampus; and a 3.4 kb transcript expressed in fetal liver, leukocytes and thymus. CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions. At least two different isoforms of CRTH2 are known to exist. Single nucleotide polymorphisms in the 3' UTR of the CRTH2 gene have been associated with asthma susceptibility.Specifications
CD294 (CRTH2) | |
0.5 mg/mL | |
Flow Cytometry | |
Biotin | |
Rat | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9Y5Y4 | |
11251 | |
Primary | |
Affinity chromatography |
BM16 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
PTGDR2 | |
CD294; chemoattractant receptor homologous molecule expressed on T helper type 2 cells; chemoattractant receptor-homologous molecule expressed on TH2 cells; CRTH2; DL1R; DP2; G protein-coupled receptor 44; Gpr44; G-protein coupled receptor 44; Grp45; MGC130436; PGD2 receptor; prostaglandin D2 receptor 2; prostaglandin D2 receptor CRTH2; Ptgdr2; putative G-protein coupled receptor 44 | |
PTGDR2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.