Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD297 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581586
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat ART4.
This gene encodes a protein that contains a mono-ADP-ribosylation motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known.
Specifications
CD297 | |
Polyclonal | |
Unconjugated | |
Art4 | |
4432404K01Rik; ADP-ribosyltransferase 4; ADP-ribosyltransferase 4 (DO blood group); ADP-ribosyltransferase 4 (Dombrock blood group); ADP-ribosyltransferase 4 (inactive) (Dombrock blood group); ADP-ribosyltransferase C2 and C3 toxin-like 4; ART4; ARTC4; ATR4; CD297; DO; DOK1; Dombrock blood group carrier molecule; ecto-ADP-ribosyltransferase 4; mono (ADP-ribosyl)transferase; mono(ADP-ribosyl)transferase 4; mono-ADP-ribosyltransferase 4; NAD(+)--protein-arginine ADP-ribosyltransferase 4; NAD(P)(+)--arginine ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
312806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
0 | |
Art4 | |
Recombinant Rat ART4 Protein. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction