Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD297 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114881 Supplier Invitrogen™ Supplier No. PA5114881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ART4.

This gene encodes a protein that contains a mono-ADP-ribosylation motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD297
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Art4
Gene Accession No. Q93070
Gene Alias 4432404K01Rik; ADP-ribosyltransferase 4; ADP-ribosyltransferase 4 (DO blood group); ADP-ribosyltransferase 4 (Dombrock blood group); ADP-ribosyltransferase 4 (inactive) (Dombrock blood group); ADP-ribosyltransferase C2 and C3 toxin-like 4; ART4; ARTC4; ATR4; CD297; DO; DOK1; Dombrock blood group carrier molecule; ecto-ADP-ribosyltransferase 4; mono (ADP-ribosyl)transferase; mono(ADP-ribosyl)transferase 4; mono-ADP-ribosyltransferase 4; NAD(+)--protein-arginine ADP-ribosyltransferase 4; NAD(P)(+)--arginine ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 4
Gene Symbols Art4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ART4(Accession Q93070), corresponding to amino acid residues L40-Q90.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 420
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.