Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD298 Mouse, anti-Human, R718, Clone: P-3E10, BD OptiBuild™

Catalog No. BDB752282 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752282 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752282 Supplier BD Biosciences Supplier No. 752282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The P-3E10 monoclonal antibody specifically recognizes CD298, the β3 subunit of the Na+/K+ ATPase (Sodium/potassium-transporting ATPase subunit beta-3). CD298 is a ∼45-55 kDa single-pass, type II membrane protein that is encoded by ATP1B3 (ATPase Na+/K+ transporting subunit beta 3) which belongs to the P-type ATPases superfamily. CD298 is widely expressed on lymphocytes, monocytes, granulocytes, platelets, and on other hematopoietic and nonhematopoietic cells and cell lines. The Na+/K+ ATPase is an integral membrane protein complex with enzymatic activity that mediates the active transport and exchange of sodium and potassium ions across the plasma membrane. This complex is composed of α and β subunits. The α subunit is a 10-membrane-spanning, catalytic protein that contains binding sites for Na +, K + and ATP. The α subunits are associated with the smaller, regulatory glycoprotein β subunits. Upon ATP hydrolysis, the Na+/K+ ATPase transports Na+ ions out of the cell in exchange for K+ ions that are transported in. This establishes a transmembrane electrochemical gradient that is essential for osmoregulation and for the transport of various nutrients and other molecules by cells. The P-3E10 antibody can reportedly inhibit the activation and proliferation of T cells and B cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD298
Applications Flow Cytometry
Classification Monoclonal
Clone P-3E10
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.