Learn More
Invitrogen™ CD299 (DC-SIGN/L) Polyclonal Antibody

Description
Peptide sequence: LVLQLLSFML LAGVLVAILV QVSKVPSSLS QEQSEQDAIY QNLTQLKAAV Sequence homology: Human: 100%; Pig: 86%.
Specifications
Specifications
| Antigen | CD299 (DC-SIGN/L) |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | CLEC4M |
| Gene Accession No. | Q9H2X3 |
| Gene Alias | CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866 |
| Gene Symbols | CLEC4M |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.